(2-Aminopyrimidin-4-yl)methanol 98%

Supplier: Ambeed
Warning

A120351-100MG A120351-10G A120351-1G A120351-250MG A120351-5G
77258-076EA 82.4 USD
77258-076 77258-078 77258-080 77258-082 77258-084
(2-Aminopyrimidin-4-yl)methanol 98%
(2-Aminopyrimidin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.12 g/mol
MDL Number: MFCD01689391
CAS Number: 2164-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR