A-366 98%

Supplier: Ambeed
Warning

Synonyms: 5'-Methoxy-6'-[3-(1-pyrrolidinyl)propoxy]spiro[cyclobutane-1,3'-indol]-2'-amin, 5'-Methoxy-6'-[3-(1-pyrrolidinyl)propoxy]spiro[cyclobutane-1,3'-[3H]indol]-2'-amine, 5'-methoxy-6'-[3-(pyrrolidin-1-yl)propoxy]spiro[cyclobutane-1,3'-indole]-2'-amine, 5'-Methoxy-6'-(3-(pyrrolidin-1-yl)propoxy)spiro[cyclobutane-1,3'-indol]-2'-amine

A120345-10MG A120345-1MG A120345-50MG A120345-5MG
77234-064EA 47.38 USD
77234-064 77234-066 77234-068 77234-070
A-366 98%
A-366

Precaución: for Research Use Only

Formula: C₁₉H₂₇N₃O₂
MW: 329.44 g/mol
MDL Number: MFCD28133403
CAS Number: 1527503-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR