4-(Methoxymethyl)-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A120264-100MG A120264-250MG
77271-452EA 83.43 USD
77271-452 77271-454
4-(Methoxymethyl)-1H-pyrrole-2-carboxylic acid 97%
4-(Methoxymethyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD28639799
CAS Number: 1550887-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR