4-Chloroacridin-9(10H)-one 97%

Supplier: Ambeed
Warning

A120218-100MG A120218-1G A120218-250MG
77246-066EA 57.01 USD
77246-066 77246-068 77246-070
4-Chloroacridin-9(10H)-one 97%
4-Chloroacridin-9(10H)-one

Precaución: for Research Use Only

Formula: C₁₃H₈ClNO
CAS Number: 69220-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR