(S)-N-((2S,3R)-1-Amino-3-hydroxy-1-oxobutan-2-yl)-1-((S)-1-((2S,3R)-2-amino-3-hydroxybutanoyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxamide 98%

Supplier: Ambeed
Warning

A119976-10MG A119976-1MG A119976-25MG A119976-5MG
77234-052EA 409.94 USD
77234-052 77234-054 77234-056 77234-058
(S)-N-((2S,3R)-1-Amino-3-hydroxy-1-oxobutan-2-yl)-1-((S)-1-((2S,3R)-2-amino-3-hydroxybutanoyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxamide 98%
(S)-N-((2S,3R)-1-Amino-3-hydroxy-1-oxobutan-2-yl)-1-((S)-1-((2S,3R)-2-amino-3-hydroxybutanoyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₈H₃₁N₅O₆
MW: 413.47 g/mol
MDL Number: MFCD12912477
CAS Number: 117928-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR