2,6-Difluoro-3-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A119947-1G A119947-250MG A119947-5G
77246-054EA 34 USD
77246-054 77246-056 77246-058
2,6-Difluoro-3-iodobenzoic acid 98%
2,6-Difluoro-3-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂IO₂
MDL Number: MFCD11040446
CAS Number: 229178-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR