3-Methyl-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A119904-100G A119904-10G A119904-1G A119904-25G A119904-5G
77271-430EA 183 USD
77271-430 77271-432 77271-434 77271-436 77271-438 77271-440
3-Methyl-1H-1,2,4-triazole 98%
3-Methyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₃H₅N₃
MDL Number: MFCD00233931
CAS Number: 7170-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR