2-Methyl-4-phenyl-1H-imidazole 98%

Supplier: Ambeed
Warning

A119804-5G A119804-250MG
77271-426EA 278 USD
77271-426 77747-873
2-Methyl-4-phenyl-1H-imidazole 98%
2-Methyl-4-phenyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MDL Number: MFCD01646082
CAS Number: 13739-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR