4-Amino-3-methylbenzonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 4-Cyano-2-methylaniline

A119705-250MG A119705-5G
77246-048EA 9.27 USD
77246-048 77571-182
4-Amino-3-methylbenzonitrile 98%
4-Amino-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16 g/mol
MDL Number: MFCD02093969
CAS Number: 78881-21-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR