4-(3-(Pyridin-4-yl)-1H-1,2,4-triazol-5-yl)picolinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: Topiroxostat

A119678-100MG A119678-50MG A119678-1G A119678-10MG A119678-5G
77284-764EA 7.21 USD
77284-764 77284-766 77571-174 77614-938 77747-871
4-(3-(Pyridin-4-yl)-1H-1,2,4-triazol-5-yl)picolinonitrile 98%
4-(3-(Pyridin-4-yl)-1H-1,2,4-triazol-5-yl)picolinonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈N₆
MW: 248.24 g/mol
MDL Number: MFCD17167057
CAS Number: 577778-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR