6-(3-(Pyrrolidin-1-yl)propoxy)-2-(4-(3-(pyrrolidin-1-yl)propoxy)phenyl)benzo[d]oxazole dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: E6446 2HCl

A119560-10MG A119560-1MG A119560-2MG A119560-5MG A119560-50MG
77234-018EA 56.65 USD
77234-018 77234-020 77234-022 77234-024 77745-876
6-(3-(Pyrrolidin-1-yl)propoxy)-2-(4-(3-(pyrrolidin-1-yl)propoxy)phenyl)benzo[d]oxazole dihydrochloride 98%
6-(3-(Pyrrolidin-1-yl)propoxy)-2-(4-(3-(pyrrolidin-1-yl)propoxy)phenyl)benzo[d]oxazole dihydrochloride

Precaución: for Research Use Only

Formula: C₂₇H₃₇Cl₂N₃O₃
MW: 522.50 g/mol
MDL Number: MFCD29472225
CAS Number: 1345675-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR