Rosuvastatin sodium salt 97%

Supplier: Ambeed
Warning

A119544-1G A119544-5G A119544-250MG
77234-014EA 35 USD
77234-014 77234-016 77745-875
Rosuvastatin sodium salt 97%
Rosuvastatin sodium salt

Precaución: for Research Use Only

Formula: C₂₂H₂₇FN₃NaO₆S
MDL Number: MFCD18783208
CAS Number: 147098-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR