tert-Butyl 3-methyl-1H-pyrazolo[4,3-c]pyridine-1-carboxylate 97%

Supplier: Ambeed
Warning

A119527-100MG A119527-1G A119527-250MG
77271-402EA 78.28 USD
77271-402 77271-404 77271-406
tert-Butyl 3-methyl-1H-pyrazolo[4,3-c]pyridine-1-carboxylate 97%
tert-Butyl 3-methyl-1H-pyrazolo[4,3-c]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂
MW: 233.27 g/mol
MDL Number: MFCD11616892
CAS Number: 1072249-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR