3-Phenoxypyridine 98%

Supplier: Ambeed
Warning

A119484-1G A119484-5G
77284-760EA 143.17 USD
77284-760 77284-762
3-Phenoxypyridine 98%
3-Phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.195 g/mol
MDL Number: MFCD00234704
CAS Number: 2176-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR