4'-Chloropropiophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chlorophenyl)propan-1-one

A119459-100G A119459-25G A119459-500G
77284-754EA 26.78 USD
77284-754 77284-756 77284-758
4'-Chloropropiophenone 97%
4'-Chloropropiophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD00000626
CAS Number: 6285-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR