2-Bromo-5-fluoroisonicotinamide 97%

Supplier: Ambeed
Warning

A119339-100MG A119339-1G A119339-250MG A119339-5G
77234-006EA 35 USD
77234-006 77234-008 77234-010 77234-012
2-Bromo-5-fluoroisonicotinamide 97%
2-Bromo-5-fluoroisonicotinamide

Precaución: for Research Use Only

Formula: C₆H₄BrFN₂O
MW: 219.011 g/mol
MDL Number: MFCD30342404
CAS Number: 1799792-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR