4-Bromo-3-(trifluoromethyl)benzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A119217-100MG
A119217-10G
A119217-250MG
A119217-25G
A119217-5G
A119217-1G
77284-728EA
7.21
USD
77284-728
77284-730
77284-732
77284-734
77284-736
77747-868
4-Bromo-3-(trifluoromethyl)benzaldehyde 97%
4-Bromo-3-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₄BrF₃O MW: 253.0200 g/mol |
MDL Number:
MFCD08059130 CAS Number: 34328-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...