ML204 hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-(piperidin-1-yl)quinoline hydrochloride

A119058-10MG A119058-1MG A119058-50MG A119058-5MG
77258-034EA 47.38 USD
77258-034 77258-036 77258-038 77258-040
ML204 hydrochloride 98+%
ML204 hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₁₉ClN₂
MW: 262.78 g/mol
MDL Number: MFCD05662337
CAS Number: 2070015-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR