1-Methyl-2-benzimidazolinone 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydro-1-methyl-2H-benzimidazol-2-one, 1-Methyl-1H-benzo[d]imidazol-2(3H)-one

A118845-1G A118845-250MG A118845-25G A118845-5G
77233-980EA 27.99 USD
77233-980 77233-982 77233-984 77233-986
1-Methyl-2-benzimidazolinone 98%
1-Methyl-2-benzimidazolinone

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD00142654
CAS Number: 1849-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR