4,5-Dichlorothiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A118797-10G A118797-5G A118797-1G A118797-250MG
77284-694EA 318.27 USD
77284-694 77284-696 77592-310 77747-864
4,5-Dichlorothiophene-2-carboxylic acid 98%
4,5-Dichlorothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₂Cl₂O₂S
MW: 197.039 g/mol
MDL Number: MFCD07366674
CAS Number: 31166-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR