4-Fluoro-2-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A118750-100G A118750-10G A118750-25G A118750-5G
77271-380EA 72.1 USD
77271-380 77271-382 77271-384 77271-386
4-Fluoro-2-nitrobenzoic acid 97%
4-Fluoro-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
MDL Number: MFCD00024300
CAS Number: 394-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR