3-Nitro-1H-1,2,4-triazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Nitro-1,2,4-1H-triazole, 3-Nitro-1,2,4-triazole
A118742-100G
A118742-10G
A118742-25G
A118742-5G
A118742-500G
77271-378EA
109
USD
77271-378
77566-180
77566-182
77571-112
77747-863
3-Nitro-1H-1,2,4-triazole 98%
3-Nitro-1H-1,2,4-triazole
Precaución: For research use only.
Formula:
C₂H₂N₄O₂ |
MDL Number:
MFCD00009749 CAS Number: 24807-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...