2,6-Dihydroxynaphthalene-1,5-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A118673-100MG A118673-1G A118673-250MG A118673-25G A118673-5G
77258-018EA 22 USD
77258-018 77258-020 77258-022 77258-024 77258-026
2,6-Dihydroxynaphthalene-1,5-dicarbaldehyde 98%
2,6-Dihydroxynaphthalene-1,5-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈O₄
MW: 216.18 g/mol
MDL Number: MFCD24713755
CAS Number: 7235-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR