2-Amino-3-hydroxypyridin 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyridin-3-ol, 2-Amino-3-hydroxypyridine

A118555-100G A118555-25G A118555-500G
77284-688EA 25 USD
77284-688 77284-690 77284-692
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.12
Boiling Pt: 250
Melting Pt: 172
MDL Number: MFCD00006317
CAS Number: 16867-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR