Isonicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine

A118427-1000G A118427-100G A118427-25G A118427-500G
77284-680EA 71.07 USD
77284-680 77284-682 77284-684 77284-686
Isonicotinonitrile 98%
Isonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₄N₂
MW: 104.11 g/mol
MDL Number: MFCD00006417
CAS Number: 100-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR