tert-Butyl 4-(((4-(4-amino-2-fluorophenoxy)-6-methoxyquinolin-7-yl)oxy)methyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A118348-250MG A118348-1G
77258-010EA 46 USD
77258-010 77614-922
tert-Butyl 4-(((4-(4-amino-2-fluorophenoxy)-6-methoxyquinolin-7-yl)oxy)methyl)piperidine-1-carboxylate 97%
tert-Butyl 4-(((4-(4-amino-2-fluorophenoxy)-6-methoxyquinolin-7-yl)oxy)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₇H₃₂FN₃O₅
MDL Number: MFCD26386170
CAS Number: 929252-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR