4-Methylcyclohexanecarboxylic acid (cis and trans mixture) 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylcyclohexanecarboxylic acid, 4-Methyl-1-cyclohexanecarboxylic acid

A118341-10G A118341-1G A118341-25G A118341-5G
77284-672EA 39 USD
77284-672 77284-674 77284-676 77284-678
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) 97%
4-Methylcyclohexanecarboxylic acid (cis and trans mixture)

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MDL Number: MFCD00074909
CAS Number: 4331-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR