2-Methyl-9,10-anthraquinone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylanthraquinone, 2-Methylanthracene-9,10-Dione

A118330-100G A118330-10G A118330-25G A118330-500G
77271-354EA 27.81 USD
77271-354 77271-356 77271-358 77271-360
2-Methyl-9,10-anthraquinone 97%
2-Methyl-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
MDL Number: MFCD00001235
CAS Number: 84-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR