3-Pyridin-4-ylpropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Pyridyl)propionic Acid

A118301-10G A118301-1G A118301-250MG A118301-25G A118301-5G
77284-662EA 273.98 USD
77284-662 77284-664 77284-666 77284-668 77284-670
3-Pyridin-4-ylpropionic acid 98%
3-Pyridin-4-ylpropionic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00084846
CAS Number: 6318-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR