N-Carbobenzoxy-D-valine 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-Val-OH, Z-D-Val-OH, N-Cbz-D-valine

A118282-100G A118282-10G A118282-5G
77271-348EA 59.74 USD
77271-348 77271-350 77271-352
N-Carbobenzoxy-D-valine 98%
N-Carbobenzoxy-D-valine

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
MDL Number: MFCD00065703
CAS Number: 1685-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR