Methyl-4-methoxypicolinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methoxypyridine-2-carboxylate

A118190-100G A118190-10G A118190-1G A118190-25G A118190-5G
77246-004EA 381.1 USD
77246-004 77246-006 77246-008 77246-010 77246-012
Methyl-4-methoxypicolinate 97%
Methyl 4-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00956223
CAS Number: 29681-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR