2-Methyl-2H-1,4-benzoxazin-3(4H)-one 97%

Supplier: Ambeed
Warning

A118095-25G A118095-5G
77258-004EA 22.66 USD
77258-004 77258-006 77258-008
2-Methyl-2H-1,4-benzoxazin-3(4H)-one 97%
2-Methyl-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD04141926
CAS Number: 21744-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR