3-Acetamido-4-methyl-2-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A118079-10G A118079-1G A118079-25G A118079-5G
77271-344EA 160 USD
77271-344 77271-346 77747-858 77747-859
3-Acetamido-4-methyl-2-nitrobenzoic acid 97%
3-Acetamido-4-methyl-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C10H10N2O5
MDL Number: MFCD00755098
CAS Number: 7356-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR