2-Phenylpiperidine 98%

Supplier: Ambeed
Warning

A118058-1G A118058-250MG A118058-25G A118058-5G
77271-332EA 25.75 USD
77271-332 77271-334 77271-336 77271-338
2-Phenylpiperidine 98%
2-Phenylpiperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N
MW: 161.25 g/mol
MDL Number: MFCD00270125
CAS Number: 3466-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR