3-Bromo-4-hydroxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-4-hydroxyphenyl)methanol, 2-Bromo-4-(hydroxymethyl)phenol

A118016-1G A118016-25G A118016-5G
77257-998EA 21.63 USD
77257-998 77258-000 77258-002
3-Bromo-4-hydroxybenzyl alcohol 98%
3-Bromo-4-hydroxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
MW: 203.04 g/mol
MDL Number: MFCD00016869
CAS Number: 29922-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR