3-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A117986-10G A117986-1G A117986-250MG A117986-5G
77271-324EA 188.49 USD
77271-324 77271-326 77271-328 77271-330
3-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine 97%
3-Bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MDL Number: MFCD11100204
CAS Number: 866546-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR