11H-Indeno[1,2-b]quinoxalin-11-one oxime 98%

Supplier: Ambeed
Warning

A117837-100MG A117837-10MG A117837-25MG A117837-50MG A117837-5MG
77233-954EA 281 USD
77233-954 77233-956 77233-958 77233-960 77233-962 77233-964
11H-Indeno[1,2-b]quinoxalin-11-one oxime 98%
11H-Indeno[1,2-b]quinoxalin-11-one oxime

Precaución: for Research Use Only

Formula: C₁₅H₉N₃O
MDL Number: MFCD00612483
CAS Number: 23146-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR