2,2-Bis(3,4-dimethylphenyl)hexafluoropropane 98%

Supplier: Ambeed
Warning

A117769-100G A117769-1G A117769-25G A117769-500G
77284-628EA 62 USD
77284-628 77284-630 77284-632 77284-634
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane 98%
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane

Precaución: for Research Use Only

Formula: C₁₉H₁₈F₆
MDL Number: MFCD00042167
CAS Number: 65294-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR