2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) 98%

Supplier: Ambeed
Warning

A117602-1G A117602-250MG A117602-25G A117602-5G
77257-982EA 43 USD
77257-982 77257-984 77257-986 77257-988
2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) 98%
2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers)

Precaución: for Research Use Only

Formula: C₁₅H₂₈O
MDL Number: MFCD27976821
CAS Number: 23790-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR