Ethyl-3-chloro-5-(trifluoromethyl)picolinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-chloro-5-(trifluoromethyl)picolinate, Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

A117586-10G A117586-1G A117586-25G A117586-5G
77271-300EA 80.34 USD
77271-300 77271-302 77271-304 77271-306
Ethyl-3-chloro-5-(trifluoromethyl)picolinate 97%
Ethyl 3-chloro-5-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₉H₇ClF₃NO₂
MW: 253.60 g/mol
MDL Number: MFCD06656414
CAS Number: 128073-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR