3-Amino-3-methyl-1-pentyne hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylpent-1-yn-3-amine hydrochloride

A117563-1G A117563-250MG A117563-25G A117563-5G
77271-292EA 53.56 USD
77271-292 77271-294 77271-296 77271-298
3-Amino-3-methyl-1-pentyne hydrochloride 97%
3-Amino-3-methyl-1-pentyne hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁N·ClH
MW: 133.619 g/mol
MDL Number: MFCD09749829
CAS Number: 108575-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR