5,6-Dichloropyridin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-3-pyridinol, 2,3-Dichloro-5-hydroxypyridine

A117492-1G A117492-250MG
77245-970EA 54.59 USD
77245-970 77245-972
5,6-Dichloropyridin-3-ol 98%
5,6-Dichloropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃Cl₂NO
MW: 163.9900 g/mol
MDL Number: MFCD09835219
CAS Number: 110860-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR