(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid

A117253-10G A117253-1G
77257-976EA 214.24 USD
77257-976 77257-978
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid 95%
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.59 g/mol
MDL Number: MFCD01631471
CAS Number: 22893-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR