DL-4-Methoxymandelic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-2-(4-methoxyphenyl)acetic acid

A117155-10G A117155-1G A117155-250MG A117155-25G A117155-5G
77284-604EA 32 USD
77284-604 77284-606 77284-608 77284-610 77284-612
DL-4-Methoxymandelic acid 98%
DL-4-Methoxymandelic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00004233
CAS Number: 10502-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR