4-Chloro-N-methylpyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A117097-100MG A117097-1G A117097-250MG
77271-274EA 48.41 USD
77271-274 77271-276 77271-278
4-Chloro-N-methylpyrimidin-2-amine 97%
4-Chloro-N-methylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.5700 g/mol
MDL Number: MFCD09834973
CAS Number: 22404-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR