3-Bromo-1-ethyl-5-methyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

A117079-250MG A117079-5G
77233-946EA 76.22 USD
77233-946 77233-948
3-Bromo-1-ethyl-5-methyl-1H-pyrazole 95%
3-Bromo-1-ethyl-5-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₉BrN₂
MW: 189.05 g/mol
MDL Number: MFCD24540323
CAS Number: 1354706-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR