3,5-Difluoroaniline 98%

Supplier: Ambeed
Warning

A117058-25G A117058-5G A117058-100G A117058-1G
77284-592EA 21 USD
77284-592 77284-594 77747-850 77747-851
3,5-Difluoroaniline 98%
3,5-Difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅F₂N
MDL Number: MFCD00007763
CAS Number: 372-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR