4-Ethylphenyl isocyanate 98%

Supplier: Ambeed
Danger

A117029-1G A117029-250MG A117029-5G
77284-586EA 22 USD
77284-586 77284-588 77284-590
4-Ethylphenyl isocyanate 98%
4-Ethylphenyl isocyanate

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00013881
CAS Number: 23138-50-3
UN: 2206
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR