4-Chloro-5,6-difluoronicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-5,6-difluoropyridine-3-carboxylic acid

A116913-100MG A116913-1G A116913-250MG A116913-5G
77284-570EA 53.56 USD
77284-570 77284-572 77284-574 77284-576
4-Chloro-5,6-difluoronicotinic acid 95%
4-Chloro-5,6-difluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MW: 193.5400 g/mol
MDL Number: MFCD09031172
CAS Number: 851386-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR