3-Bromo-4-fluorobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A116912-100G
A116912-10G
A116912-25G
A116912-500G
A116912-1000G
77271-258EA
43
USD
77271-258
77271-260
77271-262
77271-264
77549-342
3-Bromo-4-fluorobenzoic acid 97%
3-Bromo-4-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrFO₂ |
MDL Number:
MFCD00042463 CAS Number: 1007-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...